Monensin monosodium salt, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

M5273-1G M5273-500MG M5273-5G
SIALM5273-1GEA 220 GBP
SIALM5273-1G SIALM5273-500MG SIALM5273-5G
Monensin monosodium salt, Sigma-Aldrich®
Monensin monosodium salt
Formula: C₃₆H₆₁NaO₁₁
Storage Temperature: Freezer
MDL Number: MFCD00077826
CAS Number: 22373-78-0
EINECS: 244-941-7
UN: 3462
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR