Inosine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

26250.02 26250.03
SERA26250.02EA 0 GBP
SERA26250.02 SERA26250.03
Inosine
Inosine
Formula: C₁₀H₁₂N₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00066770
CAS Number: 58-63-9
EINECS: 200-390-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR