Puromycin dihydrochloride
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
SERA33835.02EA
74.76
GBP
SERA33835.02
SERA33835.01
SERA33835.03
Puromycin dihydrochloride
Puromycin dihydrochloride
Formula:
C₂₂H₂₉N₇O₅·2HCl MW: 544.44 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00012691 CAS Number: 58-58-2 EINECS: 200-387-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...