3-Quinolineboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid

011058-5G 011058-1G
FLUO011058-5GEA 0 GBP
FLUO011058-5G FLUO011058-1G
3-Quinolineboronic acid
3-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.98 g/mol
MDL Number: MFCD02183527
CAS Number: 191162-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR