MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxy-3-morpholinopropane-1-sulphonic acid
M-Clarity™ quality level = MQ400
M8389-100G
M8389-250G
M8389-25G
M8389-500G
SIALM8389-100GEA
173
GBP
SIALM8389-100G
SIALM8389-250G
SIALM8389-25G
SIALM8389-500G
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid), Sigma-Aldrich®
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)
Formula:
C₇H₁₅NO₅S Storage Temperature: Ambient |
MDL Number:
MFCD00009629 CAS Number: 68399-77-9 EINECS: 269-989-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...