N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4065417.0001
4065417.0005
BACE4065417.0001EA
0
GBP
BACE4065417.0001
BACE4065417.0005
N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)
N-Dodecanoyl-N-methyl-D-glucamine (MEGA-12)
Formula:
C₁₉H₃₇NO₇ |
CAS Number:
87246-72-8 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...