(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

30710.01 30710.02 30710.03
SERA30710.01EA 0 GBP
SERA30710.01 SERA30710.02 SERA30710.03
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)
(2-Nitrophenyl) β-D-galactopyranoside
Formula: C₁₂H₁₅NO₈
Storage Temperature: Freezer
MDL Number: MFCD00063255
CAS Number: 369-07-3
EINECS: 206-716-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR