5-Bromo-2,4-dimethoxypyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7872-1G PY-7872-25G PY-7872-5G
COBBPY-7872-1GEA 0 GBP
COBBPY-7872-1G COBBPY-7872-25G COBBPY-7872-5G
5-Bromo-2,4-dimethoxypyrimidine
5-Bromo-2,4-dimethoxypyrimidine
Formula: C₆H₇BrN₂O₂
MW: 219.0400 g/mol
MDL Number: MFCD00038016
CAS Number: 56686-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR