5-Bromo-2-(cyclopropylamino)pyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-8001-1G PY-8001-5G
COBBPY-8001-1GEA 0 GBP
COBBPY-8001-1G COBBPY-8001-5G
5-Bromo-2-(cyclopropylamino)pyrimidine
5-Bromo-2-(cyclopropylamino)pyrimidine
Formula: C₇H₈BrN₃
MDL Number: MFCD03646032
CAS Number: 886366-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR