5-Bromo-2-(ethylamino)pyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7998-1G PY-7998-5G
COBBPY-7998-1GEA 0 GBP
COBBPY-7998-1G COBBPY-7998-5G
5-Bromo-2-(ethylamino)pyrimidine
5-Bromo-2-(ethylamino)pyrimidine
Formula: C₆H₈BrN₃
MDL Number: MFCD12546487
CAS Number: 4214-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR