5-Bromo-2-(diethylamino)pyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-N,N-diethylpyrimidin-2-amine

PY-1101-5G PY-1101-1G
COBBPY-1101-5GEA 0 GBP
COBBPY-1101-5G COBBPY-1101-1G
5-Bromo-2-(diethylamino)pyrimidine
5-Bromo-2-(diethylamino)pyrimidine
Formula: C₈H₁₂BrN₃
MW: 230.1050 g/mol
MDL Number: MFCD03646011
CAS Number: 433684-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR