Bacitracin, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

B0125-1250KU B0125-250KU B0125-50KU
SIALB0125-1250KUEA 400 GBP
SIALB0125-1250KU SIALB0125-250KU SIALB0125-50KU
Bacitracin, Sigma-Aldrich®
Bacitracin
Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
MW: 1421.60 g/mol
MDL Number: MFCD00062640
CAS Number: 1405-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR