Bacitracin, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

11702-25G 11702-5G
SIAL11702-25GEA 398 GBP
SIAL11702-25G SIAL11702-5G
Bacitracin, Sigma-Aldrich®
Bacitracin
Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
MW: 1421.60 g/mol
MDL Number: MFCD00062640
CAS Number: 1405-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR