1-Chloro-2,3,4-trifluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008813-1G
008813-5G
FLUO008813-1GEA
0
GBP
FLUO008813-1G
FLUO008813-5G
1-Chloro-2,3,4-trifluorobenzene
1-Chloro-2,3,4-trifluorobenzene
Formula:
C₆H₂ClF₃ MW: 166.5300 g/mol |
MDL Number:
MFCD00042222 CAS Number: 36556-42-0 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...