3-Fluoro-4-hydroxybenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009205-1G 009205-5G 009205-25G
FLUO009205-1GEA 0 GBP
FLUO009205-1G FLUO009205-5G FLUO009205-25G
3-Fluoro-4-hydroxybenzaldehyde
3-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.11 g/mol
MDL Number: MFCD00016626
CAS Number: 405-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR