5-Bromo-2-methoxyphenol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0430-5G
OT-0430-25G
COBBOT-0430-5GEA
0
GBP
COBBOT-0430-5G
COBBOT-0430-25G
5-Bromo-2-methoxyphenol
5-Bromo-2-methoxyphenol
Formula:
C₇H₇BrO₂ MW: 203.0300 g/mol |
MDL Number:
MFCD00673137 CAS Number: 37942-01-1 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...