Hydroxyethyl cellulose, M.W. 720000, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Hydroxyethyl Cellulose

M-Clarity™ quality level = MQ100

434973-1KG 434973-250G
SIAL434973-1KGEA 228 GBP
SIAL434973-1KG SIAL434973-250G
Hydroxyethyl cellulose, M.W. 720000, Sigma-Aldrich®
Hydroxyethyl cellulose
MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR