Octaethylene glycol monohexadecylether (C16E8), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74717-1G
74717-5G
SIAL74717-1GEA
193
GBP
SIAL74717-1G
SIAL74717-5G
Octaethylene glycol monohexadecylether (C16E8), Sigma-Aldrich®
Formula:
C₃₂H₆₆O₉ |
CAS Number:
5698-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...