Didecyldimethylammonium bromide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
382310-1G
382310-5G
SIAL382310-1GEA
64.6
GBP
SIAL382310-1G
SIAL382310-5G
Didecyldimethylammonium bromide, Sigma-Aldrich®
Didecyldimethylammonium bromide
Formula:
C₂₂H₄₈BrN MW: 406.54 g/mol |
MDL Number:
MFCD00012194 CAS Number: 2390-68-3 EINECS: 219-234-1 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...