Glutarimide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Piperidine-2,6-dione
178098-25G
178098-5G
SIAL178098-25GEA
332
GBP
SIAL178098-25G
SIAL178098-5G
Glutarimide, Sigma-Aldrich®
Glutarimide
Formula:
C₅H₇NO₂ MW: 113.12 g/mol |
MDL Number:
MFCD00006670 CAS Number: 1121-89-7 EINECS: 214-340-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...