7-Bromo-1-heptanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

310913-1G 310913-5G
SIAL310913-1GEA 52 GBP
SIAL310913-1G SIAL310913-5G
7-Bromo-1-heptanol, Sigma-Aldrich®
7-Bromo-1-heptanol
Formula: C₇H₁₅BrO
MW: 195.10 g/mol
MDL Number: MFCD00041708
CAS Number: 10160-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR