tert-Butylchlorodimethylsilane
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
OT-0329-25G
OT-0329-100G
OT-0329-1KG
COBBOT-0329-25GEA
0
GBP
COBBOT-0329-25G
COBBOT-0329-100G
COBBOT-0329-1KG
tert-Butylchlorodimethylsilane
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi Boiling Pt: 125 °C (1013 hPa) Melting Pt: 87…90 °C Density: 0,81 g/cm³ (20 °C) Flash Pt: 22 °C |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...