Flash chromatography columns, PuriFlash® PT C18T Bio
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PT15C18T/1250G PT15C18T/6G PT15C18T/55G PT15C18T/450G PT15C18T/35G PT15C18T/300G PT15C18T/2500G PT15C18T/175G PT15C18T/120G PT15C18T/20G PT30C18T/450G PT30C18T/35G PT30C18T/300G PT30C18T/2500G PT30C18T/20G PT30C18T/175G PT30C18T/1250G PT30C18T/120G PT30C18T/55G PT30C18T/6G
INTEPT15C18T/1250GEA 0 GBP
INTEPT15C18T/1250G INTEPT15C18T/6G INTEPT15C18T/55G INTEPT15C18T/450G INTEPT15C18T/35G INTEPT15C18T/300G INTEPT15C18T/2500G INTEPT15C18T/175G INTEPT15C18T/120G INTEPT15C18T/20G INTEPT30C18T/450G INTEPT30C18T/35G INTEPT30C18T/300G INTEPT30C18T/2500G INTEPT30C18T/20G INTEPT30C18T/175G INTEPT30C18T/1250G INTEPT30C18T/120G INTEPT30C18T/55G INTEPT30C18T/6G
Flash chromatography columns, PuriFlash® PT C18T Bio
Chromatography Columns
These flash chromatography columns with spherical silica are suitable for the purification of weak hydrophobic peptides or oligopeptides <50 kDa.
  • Purification results equivalent to a prep LC column
  • Compatible with any flash system