Flash chromatography columns, PuriFlash®, with 300 Å C18 silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PP30C18T/6G PP15C18T/1250G PP15C18T/175G PP15C18T/120G PP15C18T/6G PP15C18T/55G PP15C18T/450G PP15C18T/35G PP15C18T/300G PP15C18T/2500G PP15C18T/20G PP30C18T/450G PP30C18T/55G PP30C18T/35G PP30C18T/300G PP30C18T/2500G PP30C18T/20G PP30C18T/175G PP30C18T/1250G PP30C18T/120G
INTEPP30C18T/6GEA 55.8 GBP
INTEPP30C18T/6G INTEPP15C18T/1250G INTEPP15C18T/175G INTEPP15C18T/120G INTEPP15C18T/6G INTEPP15C18T/55G INTEPP15C18T/450G INTEPP15C18T/35G INTEPP15C18T/300G INTEPP15C18T/2500G INTEPP15C18T/20G INTEPP30C18T/450G INTEPP30C18T/55G INTEPP30C18T/35G INTEPP30C18T/300G INTEPP30C18T/2500G INTEPP30C18T/20G INTEPP30C18T/175G INTEPP30C18T/1250G INTEPP30C18T/120G
Flash chromatography columns, PuriFlash®, with 300 Å C18 silica
Chromatography Columns
These flash chromatography columns with 300 Å C18 silica are suitable for the purification of moderately hydrophobic peptides and oligopeptides <40 kD.
  • Base material is 300 Å spherical silica
  • pH stability ranges from 1,5 to 8