Flash chromatography columns, PuriFlash®, with CN silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF50CN/2500G PF50CN/175G PF50CN/1250G PF50CN/120G PF50CN/300G PF50CN/20G PF50CN/6G PF50CN/55G PF50CN/450G PF50CN/35G
INTEPF50CN/2500GEA 4860 GBP
INTEPF50CN/2500G INTEPF50CN/175G INTEPF50CN/1250G INTEPF50CN/120G INTEPF50CN/300G INTEPF50CN/20G INTEPF50CN/6G INTEPF50CN/55G INTEPF50CN/450G INTEPF50CN/35G
Flash chromatography columns, PuriFlash®, with CN silica
Chromatography Columns
These flash chromatography columns with CN silica for mid range polarity compounds between bare silica and C18.
  • Base material is high performance spherical silica
  • pH stability ranges from 1,5 to 7