Flash chromatography columns, PuriFlash®, with 200 Å C4 silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PT15C4/20G PT15C4/175G PT15C4/1250G PT15C4/120G PT15C4/300G PT30C4/120G PT15C4/6G PT15C4/55G PT15C4/450G PT30C4/6G PT30C4/55G PT30C4/450G PT30C4/35G PT30C4/300G PT30C4/2500G PT30C4/20G PT30C4/175G PT30C4/1250G PT15C4/2500G PT15C4/35G
INTEPT15C4/20GEA 225 GBP
INTEPT15C4/20G INTEPT15C4/175G INTEPT15C4/1250G INTEPT15C4/120G INTEPT15C4/300G INTEPT30C4/120G INTEPT15C4/6G INTEPT15C4/55G INTEPT15C4/450G INTEPT30C4/6G INTEPT30C4/55G INTEPT30C4/450G INTEPT30C4/35G INTEPT30C4/300G INTEPT30C4/2500G INTEPT30C4/20G INTEPT30C4/175G INTEPT30C4/1250G INTEPT15C4/2500G INTEPT15C4/35G
Flash chromatography columns, PuriFlash®, with 200 Å C4 silica
Chromatography Columns
These flash chromatography columns with 200 Å C4 silica are suitable for the purification of hydrophobic peptides and oligopeptides <60 kD.
  • Base material 200 Å spherical silica
  • pH stability ranges from 2 to 7