Flash chromatography columns, PuriFlash®, reverse phase bonded silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF30RPAQ/35G PF30RPAQ/6G PF30RPAQ/55G PF30RPAQ/300G PF30RPAQ/450G PF30RPAQ/2500G PF30RPAQ/20G PF30RPAQ/175G PF30RPAQ/1250G PF30RPAQ/120G PF15RPAQ/120G PF15RPAQ/6G PF15RPAQ/55G PF15RPAQ/450G PF15RPAQ/35G PF15RPAQ/300G PF15RPAQ/2500G PF15RPAQ/20G PF15RPAQ/1250G PF15RPAQ/175G
INTEPF30RPAQ/35GEA 108 GBP
INTEPF30RPAQ/35G INTEPF30RPAQ/6G INTEPF30RPAQ/55G INTEPF30RPAQ/300G INTEPF30RPAQ/450G INTEPF30RPAQ/2500G INTEPF30RPAQ/20G INTEPF30RPAQ/175G INTEPF30RPAQ/1250G INTEPF30RPAQ/120G INTEPF15RPAQ/120G INTEPF15RPAQ/6G INTEPF15RPAQ/55G INTEPF15RPAQ/450G INTEPF15RPAQ/35G INTEPF15RPAQ/300G INTEPF15RPAQ/2500G INTEPF15RPAQ/20G INTEPF15RPAQ/1250G INTEPF15RPAQ/175G
Flash chromatography columns, PuriFlash®, reverse phase bonded silica
Chromatography Columns
These flash chromatography columns with reverse phase bonded silica are suitable for the purification of high and mid polar compounds.
  • 100% water compatible
  • Base material is high performance spherical silica
  • pH stability ranges from 1,5 to 7,5