Flash chromatography columns, PuriFlash®, with high capacity silica
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTEPF15SIHCJP/4GEA 331 GBP
INTEPF15SIHCJP/4G INTEPF15SIHCJP/80G INTEPF50SIHC/12G INTEPF15SIHCJP/40G INTEPF50SIHCJP200G INTEPF50SIHCJP/25G INTEPF50SIHC/4G INTEPF15SIHC/25G INTEPF25SIHC/200G INTEPF15SIHC/300G INTEPF50SIHC/80G INTEPF15SIHCJP/25G INTEPF15SIHCJP200G INTEPF15SIHC/80G INTEPF50SIHC/120G INTEPF50SIHC/40G INTEPF50SIHCJP120G INTEPF15SIHCJP1600 INTEPF15SIHC/200G INTEPF15SIHC/4G INTEPF25SIHC/300G INTEPF50SIHCJP1600 INTEPF50SIHC/200G INTEPF15SIHCJP120G INTEPF50SIHCJP/4G INTEPF25SIHC/80G INTEPF15SIHC/800G INTEPF25SIHC/25G INTEPF50SIHCJP800G INTEPF15SIHC/120G INTEPF15SIHCJP800G INTEPF50SIHC/300G INTEPF50SIHCJP/12G INTEPF25SIHC/1600G INTEPF15SIHC/12G INTEPF50SIHCJP300G INTEPF15SIHC/40G INTEPF25SIHC/800G INTEPF50SIHC/25G INTEPF50SIHCJP/80G INTEPF50SIHCJP/40G INTEPF50SIHC/800G INTEPF25SIHC/120G INTEPF25SIHC/12G INTEPF15SIHC/1600G INTEPF25SIHC/40G INTEPF25SIHC/4G INTEPF15SIHCJP/12G INTEPF50SIHC/1600G INTEPF15SIHCJP300G
Flash chromatography columns, PuriFlash®, with high capacity silica
Chromatography Columns
These flash chromatography columns with high capacity silica are suitable for the purification non-ionic polar organic compounds.
  • Methanol stable, high loading capacity and high resolution
  • Base material is high performance spherical silica
  • pH stability ranges from 1,5 to 6,5