Flash chromatography columns, PuriFlash®, with high performance silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF50SIHPJP/25G PF30SIHP/4G PF30SIHPJP200G PF30SIHPJP1600 PF30SIHPJP120G PF30SIHPJP/80G PF30SIHPJP/4G PF30SIHPJP/40G PF30SIHPJP/25G PF30SIHPJP/12G PF30SIHP/800G PF30SIHP/40G PF30SIHP/300G PF30SIHP/25G PF30SIHP/200G PF30SIHP/1600G PF30SIHP/12G PF30SIHP/120G PF30SIHP/80G PF50SIHPJP/4G PF30SIHPJP800G PF50SIHPJP/12G PF50SIHP/80G PF50SIHP/800G PF50SIHP/4G PF50SIHP/40G PF50SIHP/300G PF50SIHP/25G PF50SIHP/200G PF50SIHP/1600G PF50SIHP/120G PF50SIHP/12G PF15SIHP/800G PF50SIHPJP/40G PF15SIHP/4G PF15SIHP/80G PF15SIHP/40G PF15SIHP/25G PF15SIHP/1600G PF15SIHP/200G PF15SIHP/12G PF15SIHP/120G PF50SIHPJP800G PF50SIHPJP300G PF50SIHPJP200G PF50SIHPJP1600 PF50SIHPJP120G PF50SIHPJP/80G
INTEPF50SIHPJP/25GEA 479 GBP
INTEPF50SIHPJP/25G INTEPF30SIHP/4G INTEPF30SIHPJP200G INTEPF30SIHPJP1600 INTEPF30SIHPJP120G INTEPF30SIHPJP/80G INTEPF30SIHPJP/4G INTEPF30SIHPJP/40G INTEPF30SIHPJP/25G INTEPF30SIHPJP/12G INTEPF30SIHP/800G INTEPF30SIHP/40G INTEPF30SIHP/300G INTEPF30SIHP/25G INTEPF30SIHP/200G INTEPF30SIHP/1600G INTEPF30SIHP/12G INTEPF30SIHP/120G INTEPF30SIHP/80G INTEPF50SIHPJP/4G INTEPF30SIHPJP800G INTEPF50SIHPJP/12G INTEPF50SIHP/80G INTEPF50SIHP/800G INTEPF50SIHP/4G INTEPF50SIHP/40G INTEPF50SIHP/300G INTEPF50SIHP/25G INTEPF50SIHP/200G INTEPF50SIHP/1600G INTEPF50SIHP/120G INTEPF50SIHP/12G INTEPF15SIHP/800G INTEPF50SIHPJP/40G INTEPF15SIHP/4G INTEPF15SIHP/80G INTEPF15SIHP/40G INTEPF15SIHP/25G INTEPF15SIHP/1600G INTEPF15SIHP/200G INTEPF15SIHP/12G INTEPF15SIHP/120G INTEPF50SIHPJP800G INTEPF50SIHPJP300G INTEPF50SIHPJP200G INTEPF50SIHPJP1600 INTEPF50SIHPJP120G INTEPF50SIHPJP/80G
Flash chromatography columns, PuriFlash®, with high performance silica
Chromatography Columns
These flash chromatography columns with high performance silica are suitable for the purification non-ionic polar organic compounds.
  • Methanol stable, high separation efficiency at high flow rates
  • Base material is high performance spherical silica
  • pH stability ranges from 1,5 to 6,5