Flash chromatography columns, PuriFlash®, C18 HC silica
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF30C18HC/55G PF30C18HC/35G PF15C18HC/35G PF15C18HC/6G PF15C18HC/450G PF15C18HC/300G PF15C18HC/2500 PF15C18HC/175G PF15C18HC/1250 PF15C18HC/120G PF15C18HC/55G PF30C18HC/2500 PF30C18HC/20G PF30C18HC/175G PF30C18HC/1250 PF30C18HC/120G PF30C18HC/450G PF30C18HC/300G
INTEPF30C18HC/55GEA 129 GBP
INTEPF30C18HC/55G INTEPF30C18HC/35G INTEPF15C18HC/35G INTEPF15C18HC/6G INTEPF15C18HC/450G INTEPF15C18HC/300G INTEPF15C18HC/2500 INTEPF15C18HC/175G INTEPF15C18HC/1250 INTEPF15C18HC/120G INTEPF15C18HC/55G INTEPF30C18HC/2500 INTEPF30C18HC/20G INTEPF30C18HC/175G INTEPF30C18HC/1250 INTEPF30C18HC/120G INTEPF30C18HC/450G INTEPF30C18HC/300G
Flash chromatography columns, PuriFlash®, C18 HC silica
Chromatography Columns
These flash chromatography columns, C18 HC silica are suitable for the purification of non polar and ionic compounds.
  • Base material is high capacity spherical silica
  • pH stability ranges from 1 to 7,5