Flash chromatography columns, PuriFlash®, Cellufine™ Q500
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CF90Q5001540ML CF90Q5003100ML CF90Q500/7ML CF90Q500/68ML CF90Q500/575ML CF90Q500/45ML CF90Q500/375ML CF90Q500/27ML CF90Q500/215ML CF90Q500/145ML
INTECF90Q5001540MLEA 0 GBP
INTECF90Q5001540ML INTECF90Q5003100ML INTECF90Q500/7ML INTECF90Q500/68ML INTECF90Q500/575ML INTECF90Q500/45ML INTECF90Q500/375ML INTECF90Q500/27ML INTECF90Q500/215ML INTECF90Q500/145ML
Flash chromatography columns, PuriFlash®, Cellufine™ Q500
Chromatography Columns
These flash chromatography columns have immobilized poly (e-lysine) Cellufine™ beads which are suitable for strong anion exchange, intermediate purification of proteins, peptides and enzymes up to 500 kDa.
  • pH stability from 2 to 12
  • Capacity >10 mg/ml BSA