Flash chromatography columns, PuriFlash®, Cellufine™ DEAE
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CF90A500/215ML CF90A500/145ML CF90A500/45ML CF90A500/27ML CF90A500/375ML CF90A5003100ML CF90A5001540ML CF90A500/7ML CF90A500/68ML CF90A500/575ML
INTECF90A500/215MLEA 0 GBP
INTECF90A500/215ML INTECF90A500/145ML INTECF90A500/45ML INTECF90A500/27ML INTECF90A500/375ML INTECF90A5003100ML INTECF90A5001540ML INTECF90A500/7ML INTECF90A500/68ML INTECF90A500/575ML
Flash chromatography columns, PuriFlash®, Cellufine™ DEAE
Chromatography Columns
These flash chromatography columns have immobilised poly (e-lysine) Cellufine™ beads which are suitable for weak anion exchange, intermediate purification of proteins, peptides and enzymes up to 500 kD.
  • pH stability from 1 to 13
  • Capacity >60 mg/ml BSA