Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

37960.01 37960.02
SERA37960.01EA 0 GBP
SERA37960.01 SERA37960.02
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Formula: C₉H₁₂N₂Na₃O₁₅P₃·2H₂O
MW: 586.12 g/mol
Storage Temperature: Freezer
CAS Number: 116295-90-0
EINECS: 243-347-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR