5-Fluorouridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidine-2,4(1H,3H)-dione

F5130-100MG F5130-250MG F5130-1G
SIALF5130-100MGEA 117 GBP
SIALF5130-100MG SIALF5130-250MG SIALF5130-1G
5-Fluorouridine, Sigma-Aldrich®
5-Fluorouridine
Formula: C₉H₁₁FN₂O₆
MW: 262.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036832
CAS Number: 316-46-1
EINECS: 206-260-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR