AMPSO, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid

M-Clarity™ quality level = MQ200

A6659-100G A6659-25G
SIALA6659-100GEA 269 GBP
SIALA6659-100G SIALA6659-25G
AMPSO, Sigma-Aldrich®
AMPSO
Formula: C₇H₁₇NO₅S
MW: 227.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041777
CAS Number: 68399-79-1
EINECS: 269-991-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR