6-Methoxy-3-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51348G1
AS51348G5
ACTIAS51348G1EA
0
GBP
ACTIAS51348G1
ACTIAS51348G5
6-Methoxy-3-(tributylstannyl)pyridine
6-Methoxy-3-(tributylstannyl)pyridine
Formula:
C₁₈H₃₃NOSn |
MDL Number:
MFCD01319027 CAS Number: 164014-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...