5-Methoxy-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51356G1 AS51356G5
ACTIAS51356G1EA 0 GBP
ACTIAS51356G1 ACTIAS51356G5
5-Methoxy-3-(tributylstannyl)pyridine
5-Methoxy-3-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
MDL Number: MFCD10699160
CAS Number: 1204580-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR