3-Methoxy-2-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51355G1
AS51355G5
ACTIAS51355G1EA
0
GBP
ACTIAS51355G1
ACTIAS51355G5
3-Methoxy-2-(tributylstannyl)pyridine
3-Methoxy-2-(tributylstannyl)pyridine
Formula:
C₁₈H₃₃NOSn |
MDL Number:
MFCD10699159 CAS Number: 1094072-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...