2-Methoxy-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51189G1 AS51189G5
ACTIAS51189G1EA 0 GBP
ACTIAS51189G1 ACTIAS51189G5
2-Methoxy-3-(tributylstannyl)pyridine
2-Methoxy-3-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
MDL Number: MFCD01319029
CAS Number: 223418-74-4
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR