2-Fluoro-5-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51198G1 AS51198G5
ACTIAS51198G1EA 0 GBP
ACTIAS51198G1 ACTIAS51198G5
2-Fluoro-5-(tributylstannyl)pyridine
2-Fluoro-5-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
MDL Number: MFCD07787406
CAS Number: 1025687-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR