2-Fluoro-6-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51219G1 AS51219G5
ACTIAS51219G1EA 0 GBP
ACTIAS51219G1 ACTIAS51219G5
2-Fluoro-6-(tributylstannyl)pyridine
2-Fluoro-6-(tributylstannyl)pyridine
Formula: C₁₇H₃₀FNSn
MDL Number: MFCD09025793
CAS Number: 1025744-38-0
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR