5-Bromo-2-methoxynicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4185-1G
CA-4185-5G
COBBCA-4185-1GEA
0
GBP
COBBCA-4185-1G
COBBCA-4185-5G
5-Bromo-2-methoxynicotinic acid
5-Bromo-2-methoxy-nicotinic acid
Formula:
C₇H₆BrNO₃ MW: 232.04 g/mol |
MDL Number:
MFCD06656771 CAS Number: 54916-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...