6-Methoxyisoquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0913-5G
OT-0913-1G
COBBOT-0913-5GEA
0
GBP
COBBOT-0913-5G
COBBOT-0913-1G
6-Methoxyisoquinoline
6-Methoxyisoquinoline
Formula:
C₁₀H₉NO MW: 159.1846 g/mol |
MDL Number:
MFCD04114861 CAS Number: 52986-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...