8-Chloroisoquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6019-1G
HC-6019-5G
COBBHC-6019-1GEA
0
GBP
COBBHC-6019-1G
COBBHC-6019-5G
8-Chloroisoquinoline
8-Chloroisoquinoline
Formula:
C₉H₆ClN MW: 163.6000 g/mol |
MDL Number:
MFCD07644622 CAS Number: 34784-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...