7,8-Dichloroisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6013-5G HC-6013-1G
COBBHC-6013-5GEA 0 GBP
COBBHC-6013-5G COBBHC-6013-1G
7,8-Dichloroisoquinoline
7,8-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
MDL Number: MFCD09258638
CAS Number: 61563-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR