1,3-Dichloroisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6995-5G HC-6995-1G
COBBHC-6995-5GEA 0 GBP
COBBHC-6995-5G COBBHC-6995-1G
1,3-Dichloroisoquinoline
1,3-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
MDL Number: MFCD00034750
CAS Number: 7742-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR