2-Nitro-4-(trifluoromethyl)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007945-1G 007945-5G
FLUO007945-1GEA 0 GBP
FLUO007945-1G FLUO007945-5G
2-Nitro-4-(trifluoromethyl)phenylacetonitrile
2-Nitro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₃N₂O₂
MDL Number: MFCD00045621
CAS Number: 13544-06-4
UN: 3439
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR