3-Benzyloxyphenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018227-1G 018227-5G
FLUO018227-1GEA 0 GBP
FLUO018227-1G FLUO018227-5G
3-Benzyloxyphenylacetonitrile
3-Benzyloxyphenylacetonitrile
Formula: C₁₅H₁₃NO
MDL Number: MFCD00016396
CAS Number: 20967-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR