4-(Methoxycarbonyl)phenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

010805-5G 010805-25G
FLUO010805-5GEA 0 GBP
FLUO010805-5G FLUO010805-25G
4-(Methoxycarbonyl)phenylboronic acid
4-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MDL Number: MFCD01632203
CAS Number: 99768-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR