2-(Methoxycarbonyl)phenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

M-Clarity™ quality level = MQ100

683809-1G 683809-5G
SIAL683809-1GEA 43.6 GBP
SIAL683809-1G SIAL683809-5G
2-(Methoxycarbonyl)phenylboronic acid, Sigma-Aldrich®
2-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
MDL Number: MFCD02179452
CAS Number: 374538-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR